Product Name
2,6-Diaminopimelic acid ≥97.0% (NT)
Product Number
33240
Product Brand
ALDRICH
CAS Number
583-93-7
Molecular Weight
190.20
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Powder
TITRATION (NT) HCLO4 0.1M97.0 - 103.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE