Product Name
Cyclopropane-1,1-dicarboxylic acid 97%
Product Number
343412
Product Brand
ALDRICH
CAS Number
598-10-7
Molecular Weight
130.10
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow
APPEARANCE (FORM)Powder or Crystal or Chunk(s)
TITRATION (T) NAOH 0.1M96.5 - 103.5 %
PURITY (GC AREA %)≥ 96.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD72 MONTHS