Product Name
1-Boc-piperazine 97%
Product Number
343536
Product Brand
ALDRICH
CAS Number
57260-71-6
Molecular Weight
186.25
Test
Specification
APPEARANCEWHITE TO FAINT YELLOW POWDER, CRYSTALLINE
 POWDER OR SOLID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID96.5% (MINIMUM)
 APPROVED AUGUST 1, 2001 JSB
 REVISED JULY 22, 2004. JSB/DAD2