Product Name
5-Methyl-2-pyrazinecarboxylic acid 98%
Product Number
347647
Product Brand
ALDRICH
CAS Number
5521-55-1
Molecular Weight
138.12
Test
Specification
APPEARANCE (COLOR)White to Light Brown
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M97.5 - 102.5 %
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE