Product Name
Sodium dichloroacetate 98%
Product Number
347795
Product Brand
ALDRICH
CAS Number
2156-56-1
Molecular Weight
150.92
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Powder with Chunk(s)
TITRATION (NT) HCLO4 0.1M97.5 - 102.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE