Product Name
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution 60 wt. % in H2O
Product Number
348287
Product Brand
ALDRICH
CAS Number
3327-22-8
Molecular Weight
188.10
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
 AND/OR
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE.
TITRATION58%-68% (WITH SILVER NITRATE)
 APPROVED APRIL 20, 2005 RJM