Product Name
2-Bromo-4-methylpyridine 97%
Product Number
349984
Product Brand
ALDRICH
CAS Number
4926-28-7
Molecular Weight
172.02
Test
Specification
APPEARANCE (COLOR)Colorless to Brown to Orange
APPEARANCE (FORM)Liquid
TITRATION (NT) HCLO4 0.1M96.5 - 103.5 %
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE