Product Name
O-Tritylhydroxylamine 95%
Product Number
358894
Product Brand
ALDRICH
CAS Number
31938-11-1
Molecular Weight
275.34
Test
Specification
APPEARANCE (COLOR)White to Beige
APPEARANCE (FORM)GRANULAR POWDER
PURITY (HPLC AREA %)≥ 94.0 %
CARBON CONTENT77.8 - 87.9 %
NITROGEN CONTENT4.7 - 5.4 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE