Product Name
Methyl 1-bromocyclohexanecarboxylate 97%
Product Number
361976
Product Brand
ALDRICH
CAS Number
3196-23-4
Molecular Weight
221.09
Test
Specification
APPEARANCECOLORLESS TO YELLOW LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5-103.5% (WITH AGNO3 AFTER O2 COMBUSTION)
GAS LIQUID96.5% (MINIMUM)
 APPROVED SEPTEMBER 23, 2008 RJM