Product Name
2-(3-Methyl-3-nitrobutyl)-1,3-dioxolane 97%
Product Number
362867
Product Brand
ALDRICH
CAS Number
57620-56-1
Molecular Weight
189.21
Test
Specification
Appearance (Color)Colorless to Yellow
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (GC)≥96.5 %