Product Name
1,2-Difluoro-4,5-dimethoxybenzene 90%
Product Number
369128
Product Brand
ALDRICH
CAS Number
203059-80-7
Molecular Weight
174.14
Test
Specification
Appearance (Color)Conforms to Requirements
 Colorless to Yellow or White to Yellow
Appearance (Form)Conforms to Requirements
 Liquid or Solid
Infrared spectrumConforms to Structure
Purity (GC)≥88.0 %