Product Name
N-(4-Aminophenyl)-4-methylbenzenesulfonamide 99%
Product Number
371777
Product Brand
ALDRICH
CAS Number
6380-08-1
Molecular Weight
262.33
Test
Specification
APPEARANCE (COLOR)Off White to Dark Brown
APPEARANCE (FORM)Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
TITRATION (NT) HCLO4 0.1M98.5 - 101.5 %
PURITY (HPLC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE