Product Name
N-tert-Butylisopropylamine 97%
Product Number
374148
Product Brand
ALDRICH
CAS Number
7515-80-2
Molecular Weight
115.22
Test
Specification
Appearance (Color)Colorless to Yellow
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (Titration by HClO4)96.5 - 103.5 %
Purity (GC)≥96.5 %