Product Name
2-Hexadecanone 98%
Product Number
374180
Product Brand
ALDRICH
CAS Number
18787-63-8
Molecular Weight
240.42
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)CRYSTALLINE FLAKES
PURITY (GC AREA %)≥ 97.5 %
MELTING POINT43 - 46 C
INFRARED SPECTRUMCONFORMS TO STRUCTURE