Product Name
2,8-Quinolinediol ≥99.0% (HPLC)
Product Number
37640
Product Brand
ALDRICH
CAS Number
15450-76-7
Molecular Weight
161.16
Test
Specification
APPEARANCE (COLOR)White to Light Brown
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 99.0 %
CARBON CONTENT67.08 % (THEORY)
HYDROGEN CONTENT4.38 % (THEORY)
NITROGEN CONTENT8.69 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE