Product Name
Poly(methylphenylsiloxane) viscosity 450-550 cSt 
Product Number
378496
Product Brand
ALDRICH
CAS Number
63148-58-3
Molecular Weight
Test
Specification
APPEARANCEVISCOUS COLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
VISCOSITY450 - 550 CST @ 25 DEGREES C *
 * SUPPLIER INFORMATION
 REVISED, APPROVED JANUARY 23, 2003 TJR