Product Name
Diphenyl ditelluride 98%
Product Number
384127
Product Brand
ALDRICH
CAS Number
32294-60-3
Molecular Weight
409.41
Test
Specification
APPEARANCE (COLOR)RED-ORANGE TO BROWN
APPEARANCE (FORM)Powder
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE