Product Name
1,2,4,5-Tetracyanobenzene 97%
Product Number
391980
Product Brand
ALDRICH
CAS Number
712-74-3
Molecular Weight
178.15
Test
Specification
APPEARANCEWHITE TO TAN POWDER, CRYSTALS, CRYSTALLINE
 POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 65.1%-69.8%
 NITROGEN 30.3%-32.6%
THIN-LAYERCONSISTENT WITH 97% PURITY
SOLUBILITY25MG/ML(2.5%), ACETONE; CLEAR, COLORLESS TO
 YELLOW SOLUTION
 REVISED DECEMBER 27, 2007 RJM