Product Name
6,6-Dimethylfulvene ≥95%
Product Number
40288
Product Brand
ALDRICH
CAS Number
2175-91-9
Molecular Weight
106.17
Test
Specification
APPEARANCE (COLOR)Orange to Very Dark Orange
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 95.0 %
REFRACTIVE INDEX N20/D1.546 - 1.550
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD24 MONTHS