Product Name
Aluminum tributoxide 95%
Product Number
403350
Product Brand
ALDRICH
CAS Number
3085-30-1
Molecular Weight
246.32
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Solid or Powder or Crystals or Chunks
TITRATION (KT) EDTA 0.1M10.3 - 11.6 % AL
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE