Product Name
Lead(II) tetrakis(4-cumylphenoxy)phthalocyanine Dye content 90 %
Product Number
408875
Product Brand
ALDRICH
CAS Number
91083-54-4
Molecular Weight
1560.81
Test
Specification
APPEARANCEGREEN POWDER OR CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
UV-VISIBLE SPECTRUMIN DIOXANE
 E(713 +/-2) = 135,000 (MINIMUM)
ELEMENTAL ANALYSISCARBON 62.3%-79.3%
 NITROGEN 6.3%- 8.0%
 APPROVED AUGUST 1, 2007 RJM