Product Name
Cyclohexyl methacrylate ≥97%, contains ~60 ppm monomethyl ether hydroquinone as inhibitor
Product Number
408964
Product Brand
ALDRICH
CAS Number
101-43-9
Molecular Weight
168.23
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
 OR
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID97.0% (MINIMUM)
 APPROVED NOVEMBER 15, 2006 RJM