Product Name
Piperidine ≥99.5%, purified by redistillation
Product Number
411027
Product Brand
ALDRICH
CAS Number
110-89-4
Molecular Weight
85.15
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
COLOR (IN APHA)≤ 20 APHA
PURITY (GC AREA %)≥ 99.50 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE