Product Name
2,3-Dimethyl-1-pentene ≥97.0% (GC)
Product Number
41220
Product Brand
ALDRICH
CAS Number
3404-72-6
Molecular Weight
98.19
Test
Specification
APPEARANCE (COLOR)Colorless to Very Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.0 %
DENSITY D20/40.705 - 0.709
REFRACTIVE INDEX N20/D1.402 - 1.404
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD36 MONTHS