Product Name
2,5-Di-tert-butyl-1,4-benzoquinone 99%
Product Number
419648
Product Brand
ALDRICH
CAS Number
2460-77-7
Molecular Weight
220.31
Test
Specification
APPEARANCEYELLOW POWDER OR CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID98.5% (MINIMUM)
 APPROVED AUGUST 6, 2008 RJM