Product Name
1-Fluoro-2,4-dinitrobenzene purum p.a., ≥98.0% (GC)
Product Number
42085
Product Brand
ALDRICH
CAS Number
70-34-8
Molecular Weight
186.10
Test
Specification
APPEARANCE (COLOR)Yellow to Very Dark Yellow and Green-Yellow to Very Dark Green-Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
WATER≤ 3.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
CALCIUM (ICP)≤ 50 mg/kg
CADMIUM (ICP)≤ 50 mg/kg
COBALT (ICP)≤ 50 mg/kg
COPPER (ICP)≤ 50 mg/kg
IRON (ICP)≤ 50 mg/kg
POTASSIUM (ICP)≤ 100 mg/kg
SODIUM (ICP)≤ 100 mg/kg
NICKEL (ICP)≤ 50 mg/kg
LEAD (ICP)≤ 50 mg/kg
ZINC (ICP)≤ 50 mg/kg