Product Name
α-Terpineol 90%, technical grade
Product Number
432628
Product Brand
ALDRICH
CAS Number
98-55-5
Molecular Weight
154.25
Test
Specification
Appearance (Color)Colorless or White
Appearance (Form)Solid or Liquid
Infrared spectrumConforms to Structure
GC Purity (sum of isomers)≥89.0 %A