Product Name
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol) average Mn ~2,700
Product Number
435481
Product Brand
ALDRICH
CAS Number
9003-11-6
Molecular Weight
Test
Specification
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Ethylene Glycol by Proton NMR%
 Approximately 40 Wt%
Recommended Retest Period------------------------------
 2 Years