Product Name
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol) average Mn ~3,300
Product Number
435503
Product Brand
ALDRICH
CAS Number
9003-11-6
Molecular Weight
Test
Specification
Appearance (Color)Colorless
Appearance (Form)Viscous Liquid
Infrared spectrumConforms to Structure
Ethylene Glycol by Proton NMR%
 Approximately 10 Wt%
Cloud Point22.0 - 26.0 °C
 c = 1%, H2O
Color Test0 - 90 APHA
pH5.5 - 6.7
 c = 2.5%, H2O
Water (by Karl Fischer)≤0.20 %