Product Name
(3-Glycidyloxypropyl)trimethoxysilane ≥98%
Product Number
440167
Product Brand
ALDRICH
CAS Number
2530-83-8
Molecular Weight
236.34
Test
Specification
APPEARANCE (COLOR)Colorless to Very Faint Yellow
APPEARANCE (FORM)Liquid
COLOR (IN APHA)≤ 50 APHA
PURITY (GC AREA %)≥ 98.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRATION DATE PERIOD3 YEARS