Product Name
(1S)-(+)-3-Carene 99%
Product Number
441619
Product Brand
ALDRICH
CAS Number
498-15-7
Molecular Weight
136.23
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
SPECIFIC ROTATION (20/D)17.0 ± 1.0 Degrees
CONCENTRATIONNEAT
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD72 MONTHS