Product Name
4-(1H-Imidazol-1-yl)benzaldehyde 98%
Product Number
444405
Product Brand
ALDRICH
CAS Number
10040-98-9
Molecular Weight
172.18
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Light Beige to Beige
APPEARANCE (FORM)Powder or Flake or Crystals or Crystalline Chunk(s)
PURITY (HPLC AREA %)≥ 97.5 %
CARBON CONTENT68.0 - 71.5 %
NITROGEN CONTENT15.9 - 16.7 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE