Product Name
Tetrasodium tetrakis(μ-carbonato)dirhodate hydrate
Product Number
448494
Product Brand
ALDRICH
CAS Number
Molecular Weight
537.81 (anhydrous basis)
Test
Specification
APPEARANCEBLUE TO DARK PURPLE POWDER OR CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GRAVIMETRIC ASSAY36.5%-40.0% (DRIED BASIS) (AFTER REDUCTION
 WITH MAGNESIUM)
TITRATION1.6% H2O (MINIMUM)
 APPROVED OCTOBER 9, 2008 RJM