Product Name
3-Cyclohexene-1-carboxylic acid 97%
Product Number
453757
Product Brand
ALDRICH
CAS Number
4771-80-6
Molecular Weight
126.15
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
TITRATION (T) NAOH 0.1M96.5 - 103.5 %
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE