Product Name
Diheptyl phthalate 97%
Product Number
454818
Product Brand
ALDRICH
CAS Number
3648-21-3
Molecular Weight
362.50
Test
Specification
APPEARANCECOLORLESS TO YELLOW LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE
HIGH PRESSURE LIQUID96.5% (MINIMUM)
 APPROVED OCTOBER 6, 2003 JSB