Product Name
(R)-(+)-N,N-Dimethyl-1-(1-naphthyl)ethylamine 96%
Product Number
461490
Product Brand
ALDRICH
CAS Number
119392-95-9
Molecular Weight
199.29
Test
Specification
APPEARANCECOLORLESS TO BROWN LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
OPTICAL ROTATION+43(+/- 2) DEGREES (C=2%, METHANOL)
GAS LIQUID96.5% (MINIMUM)
 APPROVED OCTOBER 17, 2007 RJM