Product Name
4-[2-[5-[4-(Diethylamino)phenyl]-4,5-dihydro-1-phenyl-1H-pyrazol-3-yl]ethenyl]-N,N-diethylaniline
Product Number
462462
Product Brand
ALDRICH
CAS Number
57609-72-0
Molecular Weight
466.66
Test
Specification
APPEARANCEYELLOW POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
UV-VISIBLE SPECTRUMIN METHANOL
 REPORT RESULTS
ELEMENTAL ANALYSISCARBON 77.0%-82.6%
 NITROGEN 11.6%-12.4%
 APPROVED APRIL 5, 2006 RJM