Product Name
2-Octyne 97%
Product Number
484644
Product Brand
ALDRICH
CAS Number
2809-67-8
Molecular Weight
110.20
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID96.5% (MINIMUM)
 APPROVED OCTOBER 30, 2008 RJM