Product Name
Ethylbenzene solution NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Product Number
487104
Product Brand
ALDRICH
CAS Number
100-41-4
Molecular Weight
106.17
Test
Specification
APPEARANCECOLORLESS LIQUID
MISCELLANEOUS ASSAYS99.75 ATOM % D (MINIMUM)
 0.100% +/- 0.002% ETHYLBENZENE
 0.010% +/- 0.001% TMS
 2.00 +/- 0.12 INCHES TUBE FILL HEIGHT
 APPROVED MAY 22, 2008 RJM