Product Name
N,N-Dimethyl-13C2-formamide 99 atom % 13C
Product Number
493058
Product Brand
ALDRICH
CAS Number
117880-10-1
Molecular Weight
75.08
Test
Specification
APPEARANCECOLORLESS LIQUID
CARBON 13 AND PROTONCONFORMS TO STRUCTURE.
ISOTOPIC ASSAY99.0% ATOM % 13C (MINIMUM)
GAS LIQUID99.0% (MAXIMUM)
TITRATION0.5% H2O (MAXIMUM)
 APPROVED JULY 10, 2007 RJM