Product Name
4-Methyl-1-naphthaldehyde 97%
Product Number
526487
Product Brand
ALDRICH
CAS Number
33738-48-6
Molecular Weight
170.21
Test
Specification
Appearance (Color)White to Yellow
Appearance (Form)Conforms to Requirements
 Solid, Powder or Crystals
Infrared spectrumConforms to Structure
Purity (GC)≥96.5 %