Product Name
1-Ethyl-3-methylimidazolium chloride dry, ≥98.0% (T)
Product Number
53096
Product Brand
ALDRICH
CAS Number
65039-09-0
Molecular Weight
146.62
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow
APPEARANCE (FORM)Powder or Crystals
ARGENTOMETR. TITRATION≥ 98.0 %
PURITY (HPLC AREA %)≥ 98.0 %
WATER (COULOMETR.)≤ 800 ppm
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CYCLIC VOLTAMMOGRAMcorresponds to standard cyclovoltagramm
RECOMMENDED RETEST PERIOD28 MONTHS