Product Name
(R)-(+)-3-Aminopyrrolidine 98%
Product Number
540781
Product Brand
ALDRICH
CAS Number
116183-82-5
Molecular Weight
86.14
Test
Specification
Appearance (Color)Conforms to Requirements
 Colorless to Orange to Brown
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (Titration by HClO4)97.5 - 102.5 %
Optical Rotation13.0 - 23.0 deg
 c = 1%; Ethanol
Purity (GC)≥97.5 %