Product Name
4-Methylhexanoic acid 97%
Product Number
559016
Product Brand
ALDRICH
CAS Number
1561-11-1
Molecular Weight
130.18
Test
Specification
Appearance (Color)Colorless to Yellow
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (Titration by NaOH)96.5 - 103.5 %
GC (area %)≥96.5 %