Product Name
(−)-(18-Crown-6)-2,3,11,12-tetracarboxylic acid
Product Number
560111
Product Brand
ALDRICH
CAS Number
73891-15-3
Molecular Weight
440.35
Test
Specification
Appearance (Color)White
Appearance (Form)Conforms to Requirements
 Powder or Crystals
Proton NMR spectrumConforms to Structure
Carbon40.0 - 46.0 %
Optical Rotation-67.0 - -63.0 deg
 c = 1%; Methanol