Product Name
2-[(Trimethylsilyl)ethynyl]benzaldehyde 97%
Product Number
590614
Product Brand
ALDRICH
CAS Number
77123-58-1
Molecular Weight
202.32
Test
Specification
Appearance (Color)White to Beige
Appearance (Form)Conforms to Requirements
 Powder or Crystals
Infrared spectrumConforms to Structure
Titration by Hydroxylamine96.5 - 103.5 %
Purity (GC)≥96.5 %