Product Name
Magnesium L-lactate hydrate ≥95.0% (calc. based on dry substance, KT), ~2 mol/mol water
Product Number
63097
Product Brand
ALDRICH
CAS Number
1220086-24-7
Molecular Weight
202.45 (anhydrous basis)
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder
TITRATION (KT) EDTA 0.1M95.0 - 102.0 %
REMARKS ON TITRATIONCALCULATED ON DRY SUBSTANCE
SPECIFIC ROTATION (20/D)-8.5 ± 1.0 Degrees
CONCENTRATIONC=2 IN WATER
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.4G IN 10ML WATER
LOSS ON DRYING14 - 16 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE