Product Name
2,4-Dimethoxypyrimidine 97%
Product Number
635162
Product Brand
ALDRICH
CAS Number
3551-55-1
Molecular Weight
140.14
Test
Specification
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (Titration by HCL04)96.5 - 103.5 %
Purity (GC)≥96.5 %