Product Name
2-Methylallylamine hydrochloride
Product Number
639087
Product Brand
ALDRICH
CAS Number
28148-54-1
Molecular Weight
107.58
Test
Specification
Appearance (Color)White to Orange
Appearance (Form)Conforms to Requirement
 Powder or Crystals
Infrared spectrumConforms to Structure
Purity (Titration by HCL04)90.0 - 110.0 %
GC (area %)≥90.0 %