Product Name
3-(Methylthio)propylamine 97%
Product Number
639095
Product Brand
ALDRICH
CAS Number
4104-45-4
Molecular Weight
105.20
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow and Colorless to Green-Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE